missing translation for 'onlineSavingsMsg'
Learn More

7-[4-[4-(2,3-Dichlorophenyl)-1-oxidopiperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one (Aripiprazole N-Oxide), Mikromol™

Product Code. 17952126
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17952126

Brand: Mikromol MM0961.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-[4-[4-(2,3-Dichlorophenyl)-1-oxidopiperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one (Aripiprazole N-Oxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aripiprazole N-Oxide
CAS 573691-09-5
Analyte or Component Names Aripiprazole N-Oxide
Shipping Condition Room Temperature
Molecular Formula C23 H27 Cl2 N3 O3
Certifications/Compliance ISO 9001
Synonym 2(1H)-Quinolinone, 7-[4-[4-(2,3-dichlorophenyl)-1-oxido-1-piperazinyl]butoxy]-3,4-dihydro-, Aripiprazole N-Oxide, Aripiprazole Related Compound F (USP), Aripiprazole Imp. F (EP), Aripiprazole USP Related Compound F, Aripiprazole USP RC F, Aripiprazole Related Compound F, Aripiprazole Impurity F
InChI Formula InChI=1S/C23H27Cl2N3O3/c24-19-4-3-5-21(23(19)25)27-10-13-28(30,14-11-27)12-1-2-15-31-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)
SMILES [O-][N+]1(CCCCOc2ccc3CCC(=O)Nc3c2)CCN(CC1)c4cccc(Cl)c4Cl
IUPAC Name 7-[4-[4-(2,3-dichlorophenyl)-1-oxidopiperazin-1-ium-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 464.38
Quantity 25 mg
Formula Weight 463.1429 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.