Learn More
7,8-Didehydro Naloxone, TRC
Specifications
Specifications
| Chemical Name or Material | 7,8-Didehydronaloxone |
| CAS | 26451-92-3 |
| Molecular Formula | C19 H19 N O4 |
| Quantity | 25 mg |
| Synonym | Morphinan-6-one, 7,8-didehydro-4,5-epoxy-3,14-dihydroxy-17-(2-propen-1-yl)-, (5α)-, Morphinan-6-one, 17-allyl-7,8-didehydro-4,5α-epoxy-3,14-dihydroxy- (8CI), (5α)-7,8-Didehydro-4,5-epoxy-3,14-dihydroxy-17-(2-propen-1-yl)morphinan-6-one, 7,8-Didehydronaloxone, Naloxone Hydrochloride Dihydrate Imp. D (EP) |
| InChI Formula | InChI=1S/C19H19NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-6,14,17,21,23H,1,7-10H2/t14-,17+,18+,19-/m1/s1 |
| SMILES | Oc1ccc2C[C@H]3N(CC=C)CC[C@@]45[C@@H](Oc1c24)C(=O)C=C[C@@]35O |
| IUPAC Name | (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,7a,13-tetrahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one |
| Molecular Weight (g/mol) | 325.36 |
| Formula Weight | 325.1314 |