missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-3-[(5-methyl-1,3,4-thiadiazol-2-ylthio)methyl]-3-cephem-4-carboxylic Acid, TRC

Product Code. 30133538
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133538 100 mg 100mg
30010904 10 mg 10mg
30006741 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133538 Supplier Toronto Research Chemicals Supplier No. TRCA618190100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30246-33-4
Molecular Formula C11 H12 N4 O3 S3
Molecular Weight (g/mol) 344.43
Synonym (6R,7R)-7-Amino-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-amino-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo- (8CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-amino-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-, (6R-trans)-, (6R-trans)-7-Amino-3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Amino-3-(5-methyl-1,3,4-thiadiazol-2-ylthiomethyl)-δ-3-cephem-4-carboxylic acid, 7-Amino-3-[(2-methyl-1,3,4-thiadiazol-5-ylthio)methyl]-3-cephem-4-carboxylic acid, 7-Amino-3-[(5-methyl-1,3,4-thiadiazol-2-ylthio)methyl]-3-cephem-4-carboxylic acid, 7-ZACA
IUPAC Name (6R,7R)-7-amino-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](N)C3=O)C(=O)O)s1

Specifications

Analyte or Component Names 7-Amino-3-[(5-methyl-1,3,4-thiadiazol-2-ylthio)methyl]-3-cephem-4-carboxylic Acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H12N4O3S3/c1-4-13-14-11(21-4)20-3-5-2-19-9-6(12)8(16)15(9)7(5)10(17)18/h6,9H,2-3,12H2,1H3,(H,17,18)/t6-,9-/m1/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 344.0072
Chemical Name or Material 7-Amino-3-[(5-methyl-1,3,4-thiadiazol-2-ylthio)methyl]-3-cephem-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.