missing translation for 'onlineSavingsMsg'
Learn More

7-(Benzyloxy)-1-(4-((2-oxo-1,2-dihydroquinolin-7-yl)oxy)butyl)quinolin-2(1H)-one, TRC

Product Code. 30347138
Change view
Click to view available options
Quantity:
5 mg
50 mg
250 mg
Unit Size:
250mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347138 250 mg 250mg
30335114 50 mg 50mg
30336811 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30347138 Supplier Toronto Research Chemicals Supplier No. TRCB291615250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-(Benzyloxy)-1-(4-((2-oxo-1,2-dihydroquinolin-7-yl)oxy)butyl)quinolin-2(1H)-one
Analyte or Component Names 7-(Benzyloxy)-1-(4-((2-oxo-1,2-dihydroquinolin-7-yl)oxy)butyl)quinolin-2(1H)-one
Shipping Condition Room Temperature
Molecular Formula C29 H26 N2 O4
InChI Formula InChI=1S/C29H26N2O4/c32-28-14-10-22-8-12-24(18-26(22)30-28)34-17-5-4-16-31-27-19-25(13-9-23(27)11-15-29(31)33)35-20-21-6-2-1-3-7-21/h1-3,6-15,18-19H,4-5,16-17,20H2,(H,30,32)
Solution Type Neat
SMILES O=C1Nc2cc(OCCCCN3C(=O)C=Cc4ccc(OCc5ccccc5)cc34)ccc2C=C1
IUPAC Name 1-[4-[(2-oxo-1H-quinolin-7-yl)oxy]butyl]-7-phenylmethoxyquinolin-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 466.528
Quantity 250 mg
Formula Weight 466.189
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.