missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloro-6-nitro-4(3H)-quinazolinone, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Specifications
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: .. Other solubilities: slightly soluble in dmf |
Formula Weight | 225.59 |
Percent Purity | 97% |
Chemical Name or Material | 7-Chloro-6-nitro-4(3H)-quinazolinone |
Safety and Handling
Product Identifier
- 7-Chloro-6-nitroquinazolin-4-ol
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'