missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroquinoline, 97%, Thermo Scientific™

Catalog Number 15545916
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
612-61-3
C9H6ClN
163.61
QNGUPQRODVPRDC-UHFFFAOYSA-N
quinoline, 7-chloro, 7-chloro-quinoline, pubchem19899
521963
7-chloroquinoline
C1=CC2=C(C=C(C=C2)Cl)N=C1
This item is not returnable. View return policy
612-61-3
C9H6ClN
163.61
QNGUPQRODVPRDC-UHFFFAOYSA-N
quinoline, 7-chloro, 7-chloro-quinoline, pubchem19899
521963
7-chloroquinoline
C1=CC2=C(C=C(C=C2)Cl)N=C1
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 163.61
Percent Purity 97%
Chemical Name or Material 7-Chloroquinoline
compliance-icons
Product Identifier
  • 7-Chloroquinoline, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'