Learn More
Kartell™ Flask, Erlenmeyer, graduated polypropylene 24/29 500mL
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
Brand:  Kartell™ 0146300
Product Code. 10102431
  • € 46.25 / Pack of 8
Only null left
Estimated Shipment: 23-02-2026
Add to basket
Description
Description
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
  • For general laboratory use
  • Provided with standardized neck
  • Autoclavable
  • Suitable for foodstuff