Learn More
7-Hydroxymemantine Hydrochloride, Mikromol
Product Code. 30459018
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459018 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459018 Supplier Mikromol Supplier No. MM1045.120025
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-Hydroxymemantine Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-Hydroxy Memantine Hydrochloride
CAS 356572-08-2
Analyte or Component Names 7-Hydroxy Memantine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H21 N O . Cl H
Certifications/Compliance ISO 9001
Synonym 3-Amino-5,7-dimethyltricyclo[3.3.1.13,7]decan-1-ol Hydrochloride (1:1), 1-Amino-3,5-dimethyl-7-hydroxyadamantane hydrochloride
InChI Formula InChI=1S/C12H21NO.ClH/c1-9-3-10(2)5-11(13,4-9)8-12(14,6-9)7-10;/h14H,3-8,13H2,1-2H3;1H/t9-,10+,11+,12-;
Solution Type Neat
SMILES Cl.C[C@]12C[C@]3(C)C[C@@](N)(C1)C[C@@](O)(C2)C3
IUPAC Name (5S,7R)-3-amino-5,7-dimethyladamantan-1-ol;hydrochloride
Molecular Weight (g/mol) 231.76
Quantity 25 mg
Formula Weight 231.139 g/mol