missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Ketocholesterol Antibody (3F7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59357JF646
This item is not returnable.
View return policy
Description
7-Ketocholesterol Monoclonal antibody specifically detects 7-Ketocholesterol in Nonspecies specific samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| 7-Ketocholesterol | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 |
| ELISA, Immunofluorescence | |
| 3F7 | |
| ELISA, Immunocytochemistry/ Immunofluorescence | |
| 7KC, 7-KC, 7KC-Modified KLH | |
| Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 0.1 mL | |
| Cancer | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction