Learn More
Invitrogen™ 7-Ketocholesterol Monoclonal Antibody (7E1), APC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545793
Product Code. 17857162
  • € 494.00 / 100µg
Description
A 1:1,000 dilution of MA5-45793 was sufficient for detection of 7-Ketocholesterol in 2 μg of 7-ketocholesterol conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for 7-ketocholesterol modified proteins.
7-Ketocholesterol is a major oxidation product of cholesterol found in human atherosclerotic plaque and is more atherogenic than cholesterol in some animal studies. 7-Ketocholesterol can inhibit cholesterol 7alpha-hydroxylase, the rate-limiting step in bile acid biosynthesis, as well as strongly inhibiting HMG-CoA reductase, the rate-limiting enzyme in cholesterol biosynthesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7-Ketocholesterol
Monoclonal
1 mg/mL
2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG2b
ELISA, Western Blot, Immunocytochemistry
7E1
APC
7KC; 7-KC; 7KC-Modified KLH
Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μg
Primary
4°C
Liquid
Chemical