missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Catalog Number 15497538
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
69373-37-1
C8H8N2O2S
196.22
MFCD23379852
LCOAYUPAESYAHF-UHFFFAOYSA-N
7-nitro-3,4-dihydro-2h-benzo b 1,4 thiazine
20496121
7-nitro-3,4-dihydro-2H-1,4-benzothiazine
[O-][N+](=O)C1=CC2=C(NCCS2)C=C1
This item is not returnable. View return policy
69373-37-1
C8H8N2O2S
196.22
MFCD23379852
LCOAYUPAESYAHF-UHFFFAOYSA-N
7-nitro-3,4-dihydro-2h-benzo b 1,4 thiazine
20496121
7-nitro-3,4-dihydro-2H-1,4-benzothiazine
[O-][N+](=O)C1=CC2=C(NCCS2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 7-Nitro-3,4-dihydro-2H-1,4-benzothiazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'