missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 70 kDa TRITC-dextran Red Fluorophore
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Description
- Product Overview: TRITC-dextrans are dextran molecules covalently labeled with tetramethylrhodamine isothiocyanate (TRITC), offering bright red-orange fluorescence for permeability studies, molecular transport tracking, and live-cell imaging applications. Available in a wide range of molecular weights up to 2000 kDa, TRITC-dextrans are widely used in research on barrier function, vascular leakage, tissue diffusion, and cellular uptake.
- Features:
- Red-Orange Fluorescence: Excitation maximum at approximately 555 nm and emission maximum at approximately 580 nm, compatible with standard TRITC and rhodamine filter sets.
- Wide Molecular Weight Range: Available from 4 kDa up to 2000 kDa, supporting size-dependent transport and diffusion studies.
- Water-Soluble and Biocompatible: Fully dissolves in aqueous solutions, suitable for use in biological systems without significant cytotoxicity.
- Stable Labeling: TRITC conjugation ensures durable fluorescence for both dynamic tracking and endpoint assays.
- Controlled Quality: Manufactured with consistent molecular weight profiles and degrees of labeling for reproducible results.
- Benefits:
- Reliable Tracer for Barrier Studies: Quantitatively tracks transport across epithelial, endothelial, and engineered tissue barriers.
- Suitable for Live-Cell and Fixed-Tissue Imaging: Bright, stable signal supports dynamic real-time imaging and fixed-sample analysis.
- Flexible Experimental Design: Different molecular weights allow modeling of size exclusion, paracellular permeability, and tissue diffusion behavior.
- Visible Across Multiple Platforms: Detectable with standard fluorescence microscopes, flow cytometers, and plate readers.
- Applications:
- Permeability assays in epithelial and endothelial models.
- Tracking molecular diffusion in tissue engineering and microfluidic systems.
- Vascular leakage studies in preclinical models of inflammation and cancer.
- Intracellular transport and uptake research in live-cell studies.
- Fluorescent labeling for tracer studies in flow and perfusion experiments.
- Technical Specifications:
- Fluorescence: Excitation maximum about 555 nm, emission maximum about 580 nm.
- Molecular Weights: Available from 4 kDa to 2000 kDa.
- Appearance: Light-sensitive, reddish-orange dry powder, readily soluble in water.
- Storage and Stability:
- Store dry at room temperature, protected from moisture and light.
- Stable for extended periods under recommended storage conditions without significant loss of fluorescence.
- Packaging and Availability:
- Supplied as dry powder for stability and ease of use.
- Available in a range of research-grade packaging sizes to accommodate various experimental needs.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | Fluorescent Labeling Reagent |
| Molecular Weight (g/mol) | 70 kDa |
| Label Type | Other Labels or Dyes |
| Color | Red |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction