missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 70 kDa TRITC-lysine-dextran Red Fluorophore
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- Product Overview: TRITC-lysine-dextran is a water-soluble dextran modified with lysine groups and covalently labeled with tetramethylrhodamine isothiocyanate (TRITC). It offers bright red-orange fluorescence combined with enhanced conjugation and fixation capabilities through the lysine amine groups. TRITC-lysine-dextran is ideal for molecular tracking, permeability studies, and stable labeling in live-cell and fixed-cell imaging applications.
- Features:
- Red-Orange Fluorescence: Excitation maximum at approximately 555 nm and emission maximum at approximately 580 nm, compatible with TRITC filter sets.
- Lysine Modification: Provides additional amine groups, supporting enhanced conjugation and stronger fixation after labeling.
- Water-Soluble and Biocompatible: Fully soluble in aqueous systems and compatible with biological environments.
- Stable TRITC Labeling: Bright and durable fluorescence for dynamic tracking and endpoint assays.
- Available Across Multiple Sizes: Offered in various molecular weights tailored to diffusion and permeability applications.
- Benefits:
- Improved Fixation Performance: Lysine groups enhance crosslinking during paraformaldehyde fixation, preserving fluorescence for fixed-cell imaging.
- Reliable Live and Fixed Tracking: Suitable for studying real-time molecular movement and long-term fixed samples.
- Supports Surface Conjugation Studies: Facilitates binding to biomolecules and functionalized materials through available amine groups.
- Strong Signal in Complex Systems: Red-orange fluorescence ensures easy detection in tissue, cell, and fluidic models.
- Applications:
- Permeability assays across biological barriers.
- Intracellular transport and endocytosis research.
- Surface binding and conjugation experiments.
- Live-cell imaging and fixed-tissue analysis.
- Tracer studies in perfusion systems and engineered tissues.
- Technical Specifications:
- Fluorescence: Excitation maximum approximately 555 nm, emission maximum approximately 580 nm.
- Backbone: Dextran modified with lysine groups and labeled with TRITC.
- Appearance: Light-sensitive, reddish-orange dry powder, readily soluble in water.
- Storage and Stability:
- Store dry at room temperature, protected from moisture and light.
- Stable for extended periods under recommended storage conditions without significant loss of fluorescence.
- Packaging and Availability:
- Supplied as dry powder for long shelf life and convenient handling.
- Available in research-grade packaging sizes to suit different experimental needs.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Fluorescent Labeling Reagent |
| Molecular Weight (g/mol) | 70 kDa |
| Label Type | Other Labels or Dyes |
| Color | Red |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction