Learn More
Hamilton™ 700 Series Cemented Needle Custom Syringes
Click to view available options
Volume (Metric):
5 μL
10 μL
100 μL
250 μL
Unit Size:
Each
Description
- The original hand-fitted Hamilton™ syringe
- Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
- Unsurpassed syringe life
Order Info
This is a Custom Order. Please consult with your Fisher Sales Representative before ordering.
Specifications
Specifications
| Material | Borosilicate Glass |
| Diameter (Metric) Outer | 6.6 mm |
| Product Type | Syringe |
| Quantity | 1/Ea. |
| Volume (Metric) | 10 μL |
| Length (Metric) Needle | 19.1 mm |