Learn More
Abnova™ 7q33/CEN7q FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Product Type | DNA/RNA |
| Quantity | 100 μL |
| Form | Liquid |