missing translation for 'onlineSavingsMsg'
Learn More

8-Aminooctanoic acid, 99%

Product Code. p-7023614
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11443597

Brand: Thermo Scientific Alfa Aesar A17353.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1002-57-9 | C8H17NO2 | 159.23 g/mol

8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-57-9
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008245
InChI Key UQXNEWQGGVUVQA-UHFFFAOYSA-N
Synonym 8-aminocaprylic acid, octanoic acid, 8-amino, omega-aminocaprylic acid, 8-amino caprylic acid, 8-amino-octanoic acid, .omega.-aminocaprylic acid, 8-amino capylic acid, 8ac, 8-aminocapylic acid, pubchem13165
PubChem CID 66085
IUPAC Name 8-aminooctanoic acid
SMILES NCCCCCCCC(O)=O

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 159.23
Percent Purity 99%
Chemical Name or Material 8-Aminooctanoic acid
compliance-icons
Product Identifier
  • 8-Aminooctanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 8-Aminooctanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt