Learn More
8-Nitro-3,5,6,7-tetrahydro-2H-S-indacen-1-one, ≥85%, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Nitro-3,5,6,7-tetrahydro-2H-S-indacen-1-one','Technical Gra |
| CAS | 620592-45-2 |
| Molecular Formula | C12 H11 N O3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H11NO3/c14-11-5-4-9-10(11)6-7-2-1-3-8(7)12(9)13(15)16/h6H,1-5H2 |
| SMILES | [O-][N+](=O)c1c2CCCc2cc3C(=O)CCc13 |
| IUPAC Name | 4-nitro-3,5,6,7-tetrahydro-2H-s-indacen-1-one |
| Molecular Weight (g/mol) | 217.22 |
| Formula Weight | 217.0739 |