missing translation for 'onlineSavingsMsg'
Learn More

8-Pentadecanone, 98%

Product Code. 10751441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751441 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10751441 Supplier Thermo Scientific Acros Supplier No. 169790250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 818-23-5 | C15H30O | 226.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 818-23-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 40°C to 44°C
Color White to Yellow
Boiling Point 178°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C15H30O
Linear Formula CH3(CH2)6CO(CH2)6CH3
MDL Number MFCD00009555
Quantity 25 g
Beilstein 01, 717
Synonym 8-pentadecanone, caprylone, heptyl ketone, diheptyl ketone, di-n-heptyl ketone, 8-oxopentadecane, pentadecane-8-one, saturated fatty acid, acmc-1bl8g, caprylone_gurudeebansatyavani
InChI Key PQYGSSYFJIJDFK-UHFFFAOYSA-N
SMILES CCCCCCCC(=O)CCCCCCC
IUPAC Name pentadecan-8-one
Molecular Weight (g/mol) 226.40
PubChem CID 13162
Formula Weight 226.4
Percent Purity 98%
Physical Form Shiny Crystals or Flakes
Chemical Name or Material 8-Pentadecanone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.