missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 17-Propionate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 17-Propionate)
Specifications
Specifications
| Chemical Name or Material | Beclometasone 17-Propionate |
| CAS | 5534-18-9 |
| Analyte or Component Names | Beclometasone 17-Propionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H33 Cl O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Beclometasone Dipropionate Imp. H (EP), Beclometasone 17-Propionate, Beclomethasone 17-monopropionate, Beclomethasone 17-propionate, (11beta,16beta)-9-Chloro-11,21-dihydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-propionate, CCI 2382, Beclometasone Dipropionate Related Compound H, Beclometasone Dipropionate Impurity H |
| InChI Formula | InChI=1S/C25H33ClO6/c1-5-21(31)32-25(20(30)13-27)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,26)19(29)12-23(18,25)4/h8-9,11,14,17-19,27,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1 |
| SMILES | CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C)C(=O)CO |
| IUPAC Name | [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction