missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, 99%, Thermo Scientific Chemicals

Catalog Number p-98461
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13019-22-2
C10H20O
156.27
QGFSQVPRCWJZQK-UHFFFAOYSA-N
9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
25612
dec-9-en-1-ol
C=CCCCCCCCCO
This item is not returnable. View return policy
13019-22-2
C10H20O
156.27
QGFSQVPRCWJZQK-UHFFFAOYSA-N
9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
25612
dec-9-en-1-ol
C=CCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -13.0°C
Color Orange-Red to Red
Density 0.8450g/mL
Boiling Point 234.0°C to 238.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (main isomer) (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8OH
Refractive Index 1.4470 to 1.4490
Quantity 5 g
Beilstein 01,451
Specific Gravity 0.845
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Viscosity 11.4 mPa.s (20°C)
Formula Weight 156.27
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 9-Decen-1-ol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 9-Decen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'