missing translation for 'onlineSavingsMsg'
Learn More

9-Desfluoro-11-dehydroxy-9(11)-ene Triamcinolone Acetonide, TRC

Product Code. 30365868
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365868 250 mg 250mg
30357298 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365868 Supplier Toronto Research Chemicals Supplier No. TRCD289920250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5541-37-7
Molecular Formula C24 H30 O5
Molecular Weight (g/mol) 398.49
Synonym Pregna-1,4,9(11)-triene-3,20-dione, 21-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (16alpha)-, Pregna-1,4,9(11)-triene-3,20-dione, 16alpha,17,21-trihydroxy-, cyclic 16,17-acetal with acetone (7CI,8CI), 21-Hydroxy-16alpha,17alpha-(isopropylidenedioxy)pregna-1,4,9(11)triene-3,20-dione, Desonide Delta-9 (11) Analog
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)C4=CC[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C24H30O5/c1-21(2)28-20-12-18-16-6-5-14-11-15(26)7-9-22(14,3)17(16)8-10-23(18,4)24(20,29-21)19(27)13-25/h7-9,11,16,18,20,25H,5-6,10,12-13H2,1-4H3/t16-,18+,20-,22+,23+,24-/m1/s1
Formula Weight 398.2093
Chemical Name or Material (16alpha)-21-Hydroxy-16,17-((1-methylethylidene)bis(oxy))pregna-1,4,9(11)-triene-3,20-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.