Learn More
9-Fluoro-11beta-hydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl Acetate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
9-Fluoro-11beta-hydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl Acetate
Specifications
Specifications
| Chemical Name or Material | Desoxymetasone 21-Acetate |
| CAS | 1597-78-0 |
| Analyte or Component Names | Desoxymetasone 21-Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H31 F O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dexamethasone Acetate EP Impurity G, (11β,16α)-21-(Acetyloxy)-9-fluoro-11-hydroxy-16-methyl-pregna-1,4-diene-3,20-dione, 9-Fluoro-11β,21-dihydroxy-16α-methyl-pregna-1,4-diene-3,20-dione 21-Acetate, 21-Acetoxy-9-fluoro-11β-hydroxy-16α-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methylpregna-1,4-diene-11β,21-diol-3,20-dione-21-acetate |
| InChI Formula | InChI=1S/C24H31FO5/c1-13-9-18-17-6-5-15-10-16(27)7-8-23(15,4)24(17,25)20(29)11-22(18,3)21(13)19(28)12-30-14(2)26/h7-8,10,13,17-18,20-21,29H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,20+,21-,22+,23+,24+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@H]1C(=O)COC(=O)C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16R,17S)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Show More |