missing translation for 'onlineSavingsMsg'
Learn More

9-Propenyladenine, TRC

Product Code. 30011994
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011994 25 mg 25mg
30007417 100 mg 100mg
30005333 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011994 Supplier Toronto Research Chemicals Supplier No. TRCP76880025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1446486-33-4
Molecular Formula C8 H9 N5
Molecular Weight (g/mol) 175.19
Synonym 9-(1E)-1-Propen-1-yl-9H-purin-6-amine
IUPAC Name 9-[(E)-prop-1-enyl]purin-6-amine
SMILES C\C=C\n1cnc2c(N)ncnc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H9N5/c1-2-3-13-5-12-6-7(9)10-4-11-8(6)13/h2-5H,1H3,(H2,9,10,11)/b3-2+
Formula Weight 175.0858
Percent Purity >95%
Chemical Name or Material (E)-9-(Prop-1-enyl)-9H-purin-6-amine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.