Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Ideally suited for agglutination tests
- No sharp corners, which means that they can be pipetted easily and cleanly
- Suitable for +/- analyses
- Solid bottom
Specifications
Specifications
| Barcode | Alphanumeric |
| Breakapart | No |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Non Binding |
| Color | Clear |
| Product Type | Storage Microplate |
| Material | Polypropylene |
| Volume (Metric) Well | 50 to 335 μL |
| Show More |