missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosytstems™ MagMAX™ Prime Viral/Pathogen NA Isolation Kit, prefilled plate

Product Code. 30132024 Shop All Applied Biosystems Products
Click to view available options
Quantity:
96 reactions
Unit Size:
96 reactions
This item is not returnable. View return policy

Product Code. 30132024

Brand: Applied Biosytstems™ A58146PF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MagMAX Prime Viral/Pathogen NA Isolation Kit, prefilled plates, is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.

The MagMAX Prime Viral/Pathogen NA Isolation Kit, prefilled plates, is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.

Features of the MagMAX Prime Viral/Pathogen NA Isolation Kit, prefilled plates, include:
• Compatible with multiple downstream assays
• Automation-ready prefilled plates that save operators time and effort when preparing for a purification run
• Improved reagent that maximizes nucleic acid yield from routinely tested pathogens without the need for additional lysis
• Streamlined workflow accommodating various sample types and scalable sample input volume that can be run on a single plate
• Single purification protocol for various biological specimens including swabs, serum, plasma, and urine
• Designed to work with KingFisher Flex and KingFisher Apex magnetic particle processors with ability to purify 96 samples in under 60 minutes
• Sustainable product with biodegradable ingredients and reduced plastic usage
• Available accessories with optimized protocol and KingFisher script to extract nucleic acids from challenging organisms and stool sample type

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in automated magnetic bead purification techniques.

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Magnetic
Color Clear
Content And Storage

• 160 mL Binding Solution
• 300 mL Wash I Solution
• 6 mL Binding Beads
• 3 mL Proteinase K
• 36 mL Elution Buffer
• 30 μL Proteinase K dye

Store at 15–25°C.

Format Kit
Isolation Technology Beads
Sample Type Swabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Automated Protocols
Key Functions Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions 96 Reactions
Nucleic Acid Type DNA/RNA
Product Line MagMAX
Product Type Reagent
Quantity 96 reactions
Sample Volume 200–400 μL
Shipping Condition Room Temperature
Target Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.