Learn More
Uvex™ 9760 Replacement Suspension Harnesses
Replacement suspension harnesses are compatible with a variety of Pheos Safety Helmets.
POA
Specifications
Color Black
Weight (Metric) 30 g
View More Specs
Description
Description
  • For use with Uvex™ Pheos Safety Helmets
Specifications
Specifications
Black
30 g