Learn More
Uvex™ 9770 Pheos E Unventilated Safety Helmets
Lightweight, unventilated helmets in a sporty design ideal for use in the electrician sector with low voltage systems. A conventional 6-point textile strap interior allows for easy adjustment.
POA
Specifications
Certifications/Compliance | EN 397:2012, EN 50365:2002 |
---|---|
For Use With (Application) | Low Voltage Systems |
Includes | Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps |
Material | HDPE |
Model | 9770 Pheos E |
Description
- Lightweight, unventilated helmet in a sporty design
- Completely closed for use in the electrician sector with low voltage systems
- Includes an adapter system for visors, helmet caps, and helmet lamps
- Conventional 6-point textile strap interior allows for easy adjustment
- Style 9770026 is pasted with the highly reflective pheos reflex sticker set
Specifications
EN 397:2012, EN 50365:2002 | |
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps | |
9770 Pheos E | |
52 to 61 cm |
Low Voltage Systems | |
HDPE | |
Electrically Insulating Helmet | |
Not Ventilated |