Learn More
Fisherbrand™ Connecting Cable
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
Brand:  Fisherbrand™ 90603
Product Code. 10797282
  • € 58.05 / Pack of 200
Only null left
Estimated Shipment: 18-02-2026
Description
  • Connecting Cable with BNC Plug
  • For combined ion-sensitive electrode with solid membranes
  • For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
Specifications
BNC
Connecting Cable
For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug .
3 m