Learn More
A-1165442, MedChemExpress
Product Code. 16491249
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16491249
Brand: Medchem Express HY12428
MedChemExpress A-1165442 is a potent, competitive and orally available TRPV1 antagonist with an IC50 of 9 nM for human TRPV1.
A-1165442 is a potent, competitive and orally available TRPV1 antagonist with an IC50 of 9 nM for human TRPV1.
Specifications
Chemical Name or Material A-1165442
CAS 1221443-94-2
Color White
Purity Grade Notes Research
Molecular Formula C22H20ClF2N3O2
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (231.56 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=CC=CC2=C1C=C(C)N=C2)N[C@H]3C4=CC=C(Cl)C=C4OC(CF)(CF)C3
Molecular Weight (g/mol) 431.86
Formula Weight 431.86
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only