missing translation for 'onlineSavingsMsg'
Learn More

A-196, MedChemExpress

Product Code. 16414256
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414256

Brand: Medchem Express HY100201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-196 is a potent and selective inhibitor of SUV420H1 and SUV420H2 with IC50 values of 25 nM and 144 nM, respectively. A-196 inhibits SUV4-20 biochemically in a substrate-competitive manner. A-196 represents a first-in-class chemical probe of SUV4-20 to investigate the role of histone methyltransferases in genomic integrity.

A-196 is a potent and selective inhibitor of SUV420H1 and SUV420H2 with IC50 values of 25 nM and 144 nM, respectively. A-196 inhibits SUV4-20 biochemically in a substrate-competitive manner. A-196 represents a first-in-class chemical probe of SUV4-20 to investigate the role of histone methyltransferases in genomic integrity.

Specifications

Chemical Name or Material A-196
Physical Form Solid
CAS 1982372-88-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H16Cl2N4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 31 mg/mL (86.29 mM)
SMILES ClC1=C(Cl)C=C(C(C2=CC=NC=C2)=NN=C3NC4CCCC4)C3=C1
Molecular Weight (g/mol) 359.25
Formula Weight 359.25
Percent Purity 99.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.