missing translation for 'onlineSavingsMsg'
Learn More

A 438079, MedChemExpress

Product Code. 16492657
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492657 50 mg 50mg
16486568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492657 Supplier Medchem Express Supplier No. HY15488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A 438079 is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9.

A 438079 is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9.

Specifications

Chemical Name or Material A 438079
Physical Form Solid
CAS 899507-36-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H9Cl2N5
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 100 mg/mL (326.64 mM; Need ultrasonic) ∣H2O : 0.2 mg/mL (0.65 mM; Need ultrasonic)
SMILES ClC1=C(Cl)C(C2=NN=NN2CC3=CC=CN=C3)=CC=C1
Molecular Weight (g/mol) 306.15
Formula Weight 306.15
Percent Purity 99.74%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.