Learn More
A-769662, MedChemExpress
Product Code. 16498068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16498068
Brand: Medchem Express HY50662
MedChemExpress A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM.
A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM.
Specifications
Chemical Name or Material A-769662
CAS 844499-71-4
Color Light Brown
Purity Grade Notes Research
Molecular Formula C20H12N2O3S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (138.74 mM; ultrasonic and warming and heat to 80°C)
SMILES N#CC1C(C2=C(NC1=O)SC=C2C3=CC=C(C4=CC=CC=C4O)C=C3)=O
Molecular Weight (g/mol) 360.39
Formula Weight 360.39
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only