missing translation for 'onlineSavingsMsg'
Learn More
Learn More
a-Acetoxyacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Acetoxyacetic Acid |
| CAS | 13831-30-6 |
| Molecular Formula | C4 H6 O4 |
| Quantity | 1 g |
| Synonym | 2-(Acetyloxy)acetic Acid, (Acetyloxy)acetic Acid, Glycolic Acid Acetate, 2-Hydroxyacetic Acid Acetate, Acetoxyacetic Acid, Acetylglycolic Acid, NSC 72984, O-Acetylglycolic Acid |
| InChI Formula | InChI=1S/C4H6O4/c1-3(5)8-2-4(6)7/h2H2,1H3,(H,6,7) |
| SMILES | CC(=O)OCC(=O)O |
| IUPAC Name | 2-acetyloxyacetic acid |
| Molecular Weight (g/mol) | 118.09 |
| Formula Weight | 118.0266 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?