missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-Myb Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92586-0.1ml
This item is not returnable.
View return policy
Description
A-Myb Polyclonal antibody specifically detects A-Myb in Human samples. It is validated for Western Blot
Specifications
| A-Myb | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| A-MYB, AMYBMGC120059, MGC120061, Myb-like protein 1, myb-related protein A, v-myb avian myeloblastosis viral oncogene homolog-like 1, v-myb myeloblastosis viral oncogene homolog (avian)-like 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 443-692 of human MYBL1 (NP_001138227.1). PPAILRKKRKMRVGHSPGSELRDGSLNDGGNMALKHTPLKTLPFSPSQFFNTCPGNEQLNIENPSFTSTPICGQKALITTPLHKETTPKDQKENVGFRTPTIRRSILGTTPRTPTPFKNALAAQEKKYGPLKIVSQPLAFLEEDIREVLKEETGTDLFLKEEDEPAYKSCKQENTASGKKVRKSLVLDNWEKEESGTQLLTEDISDMQSNCEWETVVYGKTEDQLIMTEQARRYLSTYTATSSTSRALIL | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 4603 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction