missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A20/TNFAIP3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77024AF405
This item is not returnable.
View return policy
Description
A20/TNFAIP3 Polyclonal antibody specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| A20/TNFAIP3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human A20/TNFAIP3 . The immunogen is located within amino acids 340 - 390 of A20/TNFAIP3 Antibody. Amino Acid Squence: VQHEYKKWQENSDQARR | |
| 0.1 mL | |
| Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction | |
| 7128 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction