Learn More
Elma™ A25 Concentrated Cleaner
Description
- Suitable for metal, glass, ceramic and plastic materials
- Not suitable for aluminum, magnesium, alloys of light metals and alkali-sensitive glassware
- Removes markings, labels, greases from ground glass joints, resinified and tarry residues, lime soap deposits and prevents its redeposition, blood, saliva, protein-, bone- and tissue-residues, fats/greases and oils, polishing and grinding residues, fingerprints and dust
Specifications
Specifications
| Product Type | Lab Clean A25 Cleaner |
| pH | 12 |
| Certifications/Compliance | Annex VII A, EC 648/2004, UN 1814 |
| Composition | Amphoteric surfactants 5 to 15%, non-ionic surfactants 5 to 15%, Phosphates <5%, Polycarboxylates |
| Concentration | Ultrasonic unit 1 to 2%, dishwasher 0.5 to 1% |
| Container Type | Bottle |
| Content And Storage | 5°C to 30°C |
| For Use With (Application) | For dip and splash cleaning of instruments of labs and workshops applications |
| Volume (Metric) | 10 L |
Safety and Handling
Signal Word
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P102-Keep out of reach of children.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contain: isotridecanol,ethoxylated, C10 fatty alcohol, ethoxylated, isotridecanol, ethoxylated,potassium-hydr