Learn More
Elma™ A25 Concentrated Cleaner
Product Code. p-8001749
Click to view available options
Volume (Metric):
1 L
2.5 L
10 L
25 L
Unit Size:
1 L
10 L
2.5 L
25 L
Product Code. 11852681
Brand: Elma™ 8000087
Providing powerful cleaning power for labware and medical instruments. Elma™ A25 Concentrated Cleaner is a strongly alkaline foam-inhibited cleaner for dip and splash cleaning.
  • Suitable for metal, glass, ceramic and plastic materials
  • Not suitable for aluminum, magnesium, alloys of light metals and alkali-sensitive glassware
  • Removes markings, labels, greases from ground glass joints, resinified and tarry residues, lime soap deposits and prevents its redeposition, blood, saliva, protein-, bone- and tissue-residues, fats/greases and oils, polishing and grinding residues, fingerprints and dust
Specifications
Product Type Lab Clean A25 Cleaner
pH 12
Certifications/Compliance Annex VII A, EC 648/2004, UN 1814
Composition Amphoteric surfactants 5 to 15%, non-ionic surfactants 5 to 15%, Phosphates <5%, Polycarboxylates
Concentration Ultrasonic unit 1 to 2%, dishwasher 0.5 to 1%
Container Type Bottle
Content And Storage 5°C to 30°C
For Use With (Application) For dip and splash cleaning of instruments of labs and workshops applications
Volume (Metric) 10 L
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : isotridecanol,ethoxylated, C10 fatty alcohol, ethoxylated, isotridecanol, ethoxylated,potassium-hydr