missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A26C2/A26C3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539360
This item is not returnable.
View return policy
Description
Specifications
| A26C2/A26C3 | |
| Polyclonal | |
| Unconjugated | |
| POTEG | |
| POTE14B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23784, 404785 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6S545, Q6S5H5 | |
| POTEG, POTEH | |
| A synthetic peptide derived from the internal region of human A26C2/3 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction