missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A2BP1 Antibody (OTI4F9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71965AF350
This item is not returnable.
View return policy
Description
A2BP1 Monoclonal specifically detects A2BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| A2BP1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RBFOX1 | |
| Recombinant protein fragment corresponding to amino acids 1-210 of human A2BP1 (NP_061193) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4F9 | |
| Western Blot, Immunohistochemistry | |
| 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1 | |
| Mouse | |
| Affinity Purified | |
| Epigenetics, Neuroscience | |
| 54715.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction