missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A2M Monoclonal Antibody (K11079_7B1), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | A2M |
|---|---|
| Clone | K11079_7B1 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30194042
|
Invitrogen™
MA556214 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30208877
|
Invitrogen™
MA555834 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A2M Monoclonal antibody specifically detects A2M in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| A2M | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| A2M; A2m1; A2maa; A2MAC1; A2MD; A2mp; Alpha 2 M; Alpha-2-M; Alpha-2-M (A2M); alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; LOC100061656; Mam; S863 7; S863-7 | |
| Alpha 2 macroglobulin purified from human plasma (immunogen is full length). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| A2M |
| K11079_7B1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01023 | |
| 2 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title