Learn More
Invitrogen™ A2M Polyclonal Antibody

Description
Positive test controls include: SH-SY5Y, Mouse kidney, Mouse ovary, Rat spinal cord. The target is usually found in the following locations: Secreted. Immunogen sequence: ALAGNQDKRK EVLKSLNEEA VKKDNSVHWE RPQKPKAPVG HFYEPQAPSA EVEMTSYVLL AYLTAQPAPT SEDLTSATNI VKWITKQQNA QGGFSSTQDT VVALHALSKY GAATFTRTGK AAQVTIQSSG TFSSKFQVDN NNRLLLQQVS LPELPGEYSM KVTGEGCVYL QTSLKYNILP EKEEFPFALG VQTLPQTCDE PKAHTSFQIS LSVSYTGSRS ASNMAIVDVK MVSGFIPLKP TVKMLERSNH VSRTEVSSNH VLIYLDKVSN QTLSLFFTVL QDVPVRDLKP AIVKVYDYYE TDEFAIAEYN APCSKDLGNA.
Specifications
Specifications
| Antigen | A2M |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | A2M |
| Gene Accession No. | P01023, P06238, Q6GQT1 |
| Gene Alias | A2M; A2m1; A2maa; A2MAC1; A2MD; A2mp; Alpha 2 M; Alpha-2-M; Alpha-2-M (A2M); alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; LOC100061656; Mam; S863 7; S863-7 |
| Gene Symbols | A2M |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.