missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ A2P3 Aluminum Filter Canister

The A2P3 Aluminum Filter Canister offers high-quality, cost-effective protection against organic gases and vapors with a bp greater than 65°C. Made from aluminum with an activated charcoal fill, it is designed for low breathing resistance and limited interference with field of vision.

Brand:  Honeywell™ 1783010

Additional Details : Weight : 0.01000kg

Product Code. 12683123

  • € 42.00 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Product Type: P3 Filter Canister
  • Connection Type: Rd40
  • Material: Aluminum Alloy (Casing), Activated Charcoal (Filler), PE (End Cover)
  • Use With: Honeywell™ N7700 Mono-Filter, Half-Face, Air-Purifying Respirator (15978356)
  • Standards: EN14387, EN12941
  • EC PPE Category: 3
  • Quality Assurance: ISO 9001 / 2000
Specifications

Specifications

P3
EN14387, EN12941
Metallic Silver
Honeywell N7700 Mono-Filter, Half-Face, Air-Purifying Respirator (15978356)
EC PPE Category: 3, Quality Assurance: ISO 9001/2000
Threaded
A2P3 Aluminum Filter Canister
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers