missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A33/GPA33 Antibody (022), Alexa Fluor™ 700 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89872AF700
This item is not returnable.
View return policy
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for ELISA
Specifications
| A33/GPA33 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human A33/GPA33 (Q99795-1; Met1-Val235). | |
| 0.1 mL | |
| Cancer | |
| 10223 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 022 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction