missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A33/GPA33 Antibody (402104), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3080H
This item is not returnable.
View return policy
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| A33/GPA33 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 402104 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33 | |
| Mouse myeloma cell line NS0-derived recombinant human A33, Ile22-Val235, Accession # Q99795 | |
| 0.1 mL | |
| Cancer | |
| 10223 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction