Learn More
A33/GPA33 Antibody (4H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26549-50ul
 View more versions of this product
Product Code. 30044000
  • € 343.00 / 50µL
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A33/GPA33
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
4H4
ELISA, Immunohistochemistry 1:50-1:200
A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33
A synthesized peptide derived from Human A33/GPA33 [UniProt Q99795]
50 μL
Cancer
10223
Store at -20 to -70C. Avoid freeze-thaw cycles.