Learn More
Novus Biologicals™ A33/GPA33 Antibody (KRN330), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28143-1mg
 View more versions of this product
Product Code. 30046776
  • € 920.00 / 1mg
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A33/GPA33
Monoclonal
Unconjugated
Q99795
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
KRN330
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33
GPA33
1 mg
Cancer
10223