Learn More
Invitrogen™ A33 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598802
Product Code. 16571622
  • € 464.00 / 100µg
Description
A33 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
A33
Polyclonal
Unconjugated
Gpa33
2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33
Rabbit
Protein G
RUO
10223
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
4 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q99795
Gpa33
Recombinant Human Cell surface A33 antigen protein (22-235AA).