missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A4GNT Antibody (OTI2H9), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71905AF594
This item is not returnable.
View return policy
Description
A4GNT Monoclonal specifically detects A4GNT in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| A4GNT | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| A4GNT | |
| Recombinant protein fragment corresponding to amino acids 121-340 of human A4GNT (NP_057245) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2H9 | |
| Western Blot, Immunohistochemistry | |
| alpha-1,4-N-acetylglucosaminyltransferase | |
| Mouse | |
| 0.1 mL | |
| 51146 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction