missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A4GNT Monoclonal Antibody (OTI1B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800432S
This item is not returnable.
View return policy
Description
A4GNT Monoclonal antibody specifically detects A4GNT in Human samples. It is validated for Western BlotSpecifications
| A4GNT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NAM6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 201516 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1B10 | |
| Unconjugated | |
| ZSCAN4 | |
| A4GNT; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4 GnT; Alpha4GnT; AV080780; EGK_12072; Gm798; GnTalpha4; MGC149493 | |
| Human recombinant protein fragment corresponding to amino acids 164-433 of human ZSCAN4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction