Learn More
Tocris Bioscience™ A66
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A66 is a potent and selective PI 3-kinase p110α inhibitor (IC50 = 32 nM). Exhibits >100-fold selectivity for p110α over other PI 3-kinase isoforms. Inhibits Akt signaling and tumor growth in SK-OV-3 xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | (2S)-N1-[2-(1,1-Dimethylethyl)-4′-methyl[4,5′-bithiazol]-2′-yl]-1,2-pyrrolidinedicarboxamide |
| CAS | 1166227-08-2 |
| Quantity | 50 mg |
| Target | PI 3-Kinase Inhibitors |
| Molecular Formula | C17H23N5O2S2 |
| Formula Weight | 393.53 |
| Purity | >98% |